Anti-CD74 Rat Monoclonal Antibody [clone: 829706]

Supplier: R & D Systems
MAB7478
102886-804EA 594.37 USD
102886-804
Anti-CD74 Rat Monoclonal Antibody [clone: 829706]
Antibodies
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: 829706
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR