Anti-MINA Mouse Monoclonal Antibody [clone: 753002]

Supplier: R & D Systems
MAB7476
102886-802EA 561.11 USD
102886-802
Anti-MINA Mouse Monoclonal Antibody [clone: 753002]
Antibodies
Type: Primary
Antigen: MINA
Clonality: Monoclonal
Clone: 753002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR