Anti-PTPRU Mouse Monoclonal Antibody [clone: 764209]

Supplier: R & D Systems
MAB7475
102886-800EA 577.02 USD
102886-800
Anti-PTPRU Mouse Monoclonal Antibody [clone: 764209]
Antibodies
Type: Primary
Antigen: PTPRU
Clonality: Monoclonal
Clone: 764209
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR