Anti-LCP2 Mouse Monoclonal Antibody [clone: 776503]

Supplier: R & D Systems
MAB7474
102886-798EA 577.02 USD
102886-798
Anti-LCP2 Mouse Monoclonal Antibody [clone: 776503]
Antibodies
Simple Western

Type: Primary
Antigen: LCP2
Clonality: Monoclonal
Clone: 776503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR