Anti-SEMA4D Mouse Monoclonal Antibody [clone: 758726]

Supplier: R & D Systems
MAB74701
102886-796EA 594.37 USD
102886-796
Anti-SEMA4D Mouse Monoclonal Antibody [clone: 758726]
Antibodies
Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 758726
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR