Anti-SEMA4D Mouse Monoclonal Antibody [clone: 758734]

Supplier: R & D Systems
MAB7470
102886-794EA 584.25 USD
102886-794
Anti-SEMA4D Mouse Monoclonal Antibody [clone: 758734]
Antibodies
Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 758734
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR