Anti-S100 A6 Mouse Monoclonal Antibody [clone: 754102]

Supplier: R & D Systems
MAB7467
102887-176EA 561.11 USD
102887-176
Anti-S100 A6 Mouse Monoclonal Antibody [clone: 754102]
Antibodies
Type: Primary
Antigen: S100 A6
Clonality: Monoclonal
Clone: 754102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR