Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 182525]

Supplier: R & D Systems
MAB7461
220021-505EA 772.25 USD
220021-505
Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 182525]
Antibodies
Type: Primary
Antigen: LEFTY2
Clonality: Monoclonal
Clone: 182525
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR