Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 182515]

Supplier: R & D Systems
MAB746
220021-504EA 763.57 USD
220021-504
Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 182515]
Antibodies
Type: Primary
Antigen: LEFTY2
Clonality: Monoclonal
Clone: 182515
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR