Anti-SULT1C2 Mouse Monoclonal Antibody [clone: 844529]

Supplier: R & D Systems
MAB7458
102887-174EA 577.02 USD
102887-174
Anti-SULT1C2 Mouse Monoclonal Antibody [clone: 844529]
Antibodies
Type: Primary
Antigen: SULT1C2
Clonality: Monoclonal
Clone: 844529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR