Anti-CPM Mouse Monoclonal Antibody [clone: 848543]

Supplier: R & D Systems
MAB7457
102887-172EA 594.37 USD
102887-172
Anti-CPM Mouse Monoclonal Antibody [clone: 848543]
Antibodies
Type: Primary
Antigen: CPM
Clonality: Monoclonal
Clone: 848543
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR