Anti-Endophilin B1/B2 Mouse Monoclonal Antibody [clone: 807009]

Supplier: R & D Systems
MAB7456
102887-170EA 577.02 USD
102887-170
Anti-Endophilin B1/B2 Mouse Monoclonal Antibody [clone: 807009]
Antibodies
Simple Western

Type: Primary
Antigen Symbol:
Antigen Name: Endophilin B1/B2
Antigen Synonyms:
Clonality: Monoclonal
Clone: 807009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR