Anti-IRS1 Mouse Monoclonal Antibody [clone: 744353]

Supplier: R & D Systems
MAB7455
102887-168EA 577.02 USD
102887-168
Anti-IRS1 Mouse Monoclonal Antibody [clone: 744353]
Antibodies
Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: 744353
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR