Anti-CHST3 Mouse Monoclonal Antibody [clone: 799011]

Supplier: R & D Systems
MAB7450
102887-122EA 561.11 USD
102887-122
Anti-CHST3 Mouse Monoclonal Antibody [clone: 799011]
Antibodies
Type: Primary
Antigen: CHST3
Clonality: Monoclonal
Clone: 799011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR