Anti-EDAR Rat Monoclonal Antibody [clone: 132102]

Supplier: R & D Systems
MAB745
220021-502EA 763.57 USD
220021-502
Anti-EDAR Rat Monoclonal Antibody [clone: 132102]
Antibodies
Type: Primary
Antigen: EDAR
Clonality: Monoclonal
Clone: 132102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR