Anti-KLRB1 Mouse Monoclonal Antibody [clone: 702228]

Supplier: R & D Systems
MAB7448
102887-120EA 594.37 USD
102887-120
Anti-KLRB1 Mouse Monoclonal Antibody [clone: 702228]
Antibodies
Type: Primary
Antigen: KLRB1
Clonality: Monoclonal
Clone: 702228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR