Anti-UCN Mouse Monoclonal Antibody [clone: 749606]

Supplier: R & D Systems
MAB7447
102887-118EA 594.37 USD
102887-118
Anti-UCN Mouse Monoclonal Antibody [clone: 749606]
Antibodies
Type: Primary
Antigen: UCN
Clonality: Monoclonal
Clone: 749606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR