Anti-CD1B Mouse Monoclonal Antibody [clone: 737249]

Supplier: R & D Systems
MAB7446
102887-116EA 594.37 USD
102887-116
Anti-CD1B Mouse Monoclonal Antibody [clone: 737249]
Antibodies
Type: Primary
Antigen: CD1B
Clonality: Monoclonal
Clone: 737249
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR