Anti-TNFSF15 Rat Monoclonal Antibody [clone: 293327]

Supplier: R & D Systems
MAB7441
220021-501EA 655.11 USD
220021-501
Anti-TNFSF15 Rat Monoclonal Antibody [clone: 293327]
Antibodies
Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: 293327
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR