Anti-SH2D1A Mouse Monoclonal Antibody [clone: 782702]

Supplier: R & D Systems
MAB7440
102887-112EA 577.02 USD
102887-112
Anti-SH2D1A Mouse Monoclonal Antibody [clone: 782702]
Antibodies
Type: Primary
Antigen: SH2D1A
Clonality: Monoclonal
Clone: 782702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR