Anti-BATF3 Mouse Monoclonal Antibody [clone: 841702]

Supplier: R & D Systems
MAB7437
102887-110EA 594.37 USD
102887-110
Anti-BATF3 Mouse Monoclonal Antibody [clone: 841702]
Antibodies
Type: Primary
Antigen: BATF3
Clonality: Monoclonal
Clone: 841702
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR