Anti-CLEC14A Mouse Monoclonal Antibody [clone: 743940]

Supplier: R & D Systems
MAB7436
102887-108EA 577.02 USD
102887-108
Anti-CLEC14A Mouse Monoclonal Antibody [clone: 743940]
Antibodies
Type: Primary
Antigen: CLEC14A
Clonality: Monoclonal
Clone: 743940
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR