Anti-FLT4 Rat Monoclonal Antibody [clone: 102804]

Supplier: R & D Systems
MAB7431
220021-500EA 763.57 USD
220021-500
Anti-FLT4 Rat Monoclonal Antibody [clone: 102804]
Antibodies
Type: Primary
Antigen Symbol: FLT4
Antigen Name: FMS-like tyrosine kinase 4
Antigen Synonyms: AI323512, Chy, Flt-4, VEGFR-3, VEGFR3
Clonality: Monoclonal
Clone: 102804
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR