Anti-FLT4 Rat Monoclonal Antibody [clone: 102806]

Supplier: R & D Systems
MAB743
220021-499EA 763.57 USD
220021-499
Anti-FLT4 Rat Monoclonal Antibody [clone: 102806]
Antibodies
Type: Primary
Antigen Symbol: FLT4
Antigen Name: FMS-like tyrosine kinase 4
Antigen Synonyms: AI323512, Chy, Flt-4, VEGFR-3, VEGFR3
Clonality: Monoclonal
Clone: 102806
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR