Anti-TXNRD1 Mouse Monoclonal Antibody [clone: 489804]

Supplier: R & D Systems
MAB7428
102887-102EA 582.8 USD
102887-102
Anti-TXNRD1 Mouse Monoclonal Antibody [clone: 489804]
Antibodies
Type: Primary
Antigen: TXNRD1
Clonality: Monoclonal
Clone: 489804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR