Anti-POU3F2 Mouse Monoclonal Antibody [clone: 761340]

Supplier: R & D Systems
MAB7427
102887-100EA 561.11 USD
102887-100
Anti-POU3F2 Mouse Monoclonal Antibody [clone: 761340]
Antibodies
Type: Primary
Antigen: POU3F2
Clonality: Monoclonal
Clone: 761340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR