Anti-PLG Rat Monoclonal Antibody [clone: 120304]

Supplier: R & D Systems
MAB742
220021-498EA 763.57 USD
220021-498
Anti-PLG Rat Monoclonal Antibody [clone: 120304]
Antibodies
Type: Primary
Antigen: PLG
Clonality: Monoclonal
Clone: 120304
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR