Anti-AKT1 Mouse Monoclonal Antibody [clone: 65832]

Supplier: R & D Systems
MAB7419
102887-096EA 594.37 USD
102887-096
Anti-AKT1 Mouse Monoclonal Antibody [clone: 65832]
Antibodies
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 65832
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR