Anti-CAV1 Mouse Monoclonal Antibody [clone: 750304]

Supplier: R & D Systems
MAB7418
102887-094EA 577.02 USD
102887-094
Anti-CAV1 Mouse Monoclonal Antibody [clone: 750304]
Antibodies
Type: Primary
Antigen: CAV1
Clonality: Monoclonal
Clone: 750304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR