Anti-PUM2 Mouse Monoclonal Antibody [clone: 740610]

Supplier: R & D Systems
MAB7415
102887-092EA 577.02 USD
102887-092
Anti-PUM2 Mouse Monoclonal Antibody [clone: 740610]
Antibodies
Type: Primary
Antigen: PUM2
Clonality: Monoclonal
Clone: 740610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR