Anti-GBA Mouse Monoclonal Antibody [clone: 812201]

Supplier: R & D Systems
MAB7410
102887-090EA 655.11 USD
102887-090
Anti-GBA Mouse Monoclonal Antibody [clone: 812201]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: GBA
Antigen Name: glucosidase, beta, acid
Antigen Synonyms: GBA1, GCB, GLUC
Clonality: Monoclonal
Clone: 812201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR