Anti-CD86 Rat Monoclonal Antibody [clone: GL1]

Supplier: R & D Systems
MAB741
220021-497EA 772.25 USD
220021-497
Anti-CD86 Rat Monoclonal Antibody [clone: GL1]
Antibodies
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR