Anti-KYNU Rat Monoclonal Antibody [clone: 771312]

Supplier: R & D Systems
MAB7389
102887-088EA 577.02 USD
102887-088
Anti-KYNU Rat Monoclonal Antibody [clone: 771312]
Antibodies
Type: Primary
Antigen: KYNU
Clonality: Monoclonal
Clone: 771312
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR