Anti-ANGPT4 Rat Monoclonal Antibody [clone: 11354]

Supplier: R & D Systems
MAB738
220021-495EA 763.57 USD
220021-495
Anti-ANGPT4 Rat Monoclonal Antibody [clone: 11354]
Antibodies
Type: Primary
Antigen: ANGPT4
Clonality: Monoclonal
Clone: 11354
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR