Anti-PLCG2 Mouse Monoclonal Antibody [clone: 744757]

Supplier: R & D Systems
MAB7377
102887-086EA 594.37 USD
102887-086
Anti-PLCG2 Mouse Monoclonal Antibody [clone: 744757]
Antibodies
Type: Primary
Antigen: PLCG2
Clonality: Monoclonal
Clone: 744757
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR