Anti-AMER1 Mouse Monoclonal Antibody [clone: 729419]

Supplier: R & D Systems
MAB7374
102887-084EA 577.02 USD
102887-084
Anti-AMER1 Mouse Monoclonal Antibody [clone: 729419]
Antibodies
Type: Primary
Antigen: AMER1
Clonality: Monoclonal
Clone: 729419
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR