Anti-CCL3 Mouse Monoclonal Antibody [clone: 750521]

Supplier: R & D Systems
MAB7370
102887-082EA 561.11 USD
102887-082
Anti-CCL3 Mouse Monoclonal Antibody [clone: 750521]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 750521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR