Anti-SELP Rat Monoclonal Antibody [clone: 127933]

Supplier: R & D Systems
MAB737
220021-494EA 772.25 USD
220021-494
Anti-SELP Rat Monoclonal Antibody [clone: 127933]
Antibodies
Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: 127933
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR