Anti-LCK Mouse Monoclonal Antibody [clone: 755216]

Supplier: R & D Systems
MAB7369
102887-080EA 639.2 USD
102887-080
Anti-LCK Mouse Monoclonal Antibody [clone: 755216]
Antibodies
Type: Primary
Antigen: LCK
Clonality: Monoclonal
Clone: 755216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR