Anti-CD15 Mouse Monoclonal Antibody [clone: ICRF29-2]

Supplier: R & D Systems
MAB7368
102887-078EA 616.06 USD
102887-078
Anti-CD15 Mouse Monoclonal Antibody [clone: ICRF29-2]
Antibodies
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: ICRF29-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR