Anti-DSC1 Rat Monoclonal Antibody [clone: 772906]

Supplier: R & D Systems
MAB7367
102887-076EA 633.42 USD
102887-076
Anti-DSC1 Rat Monoclonal Antibody [clone: 772906]
Antibodies
Type: Primary
Antigen Symbol: DSC1
Antigen Name: desmocollin 1
Antigen Synonyms: 1110020A10Rik, AI507491, Dsc1a, Dsc1b
Clonality: Monoclonal
Clone: 772906
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR