Anti-IL10 Mouse Monoclonal Antibody [clone: 137706]

Supplier: R & D Systems
MAB736
220021-492EA 763.57 USD
220021-492
Anti-IL10 Mouse Monoclonal Antibody [clone: 137706]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 137706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR