Anti-CDK5R1 Mouse Monoclonal Antibody [clone: 739304]

Supplier: R & D Systems
MAB7357
102887-072EA 561.11 USD
102887-072
Anti-CDK5R1 Mouse Monoclonal Antibody [clone: 739304]
Antibodies
Type: Primary
Antigen: CDK5R1
Clonality: Monoclonal
Clone: 739304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR