Anti-IL10 Mouse Monoclonal Antibody [clone: 138111]

Supplier: R & D Systems
MAB7351
220021-490EA 772.25 USD
220021-490
Anti-IL10 Mouse Monoclonal Antibody [clone: 138111]
Antibodies
Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 138111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR