Anti-CHD7 Mouse Monoclonal Antibody [clone: 772503]

Supplier: R & D Systems
MAB7350
102887-070EA 577.02 USD
102887-070
Anti-CHD7 Mouse Monoclonal Antibody [clone: 772503]
Antibodies
Type: Primary
Antigen: CHD7
Clonality: Monoclonal
Clone: 772503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR