Anti-MST1 Mouse Monoclonal Antibody [clone: 68801]

Supplier: R & D Systems
MAB735
220021-489EA 763.57 USD
220021-489
Anti-MST1 Mouse Monoclonal Antibody [clone: 68801]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: MST1
Clonality: Monoclonal
Clone: 68801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR