Anti-TGFB2 Rat Monoclonal Antibody [clone: 771244]

Supplier: R & D Systems
MAB73461
102887-068EA 594.37 USD
102887-068
Anti-TGFB2 Rat Monoclonal Antibody [clone: 771244]
Antibodies
Type: Primary
Antigen: TGFB2
Clonality: Monoclonal
Clone: 771244
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR