Anti-LAMA4 Mouse Monoclonal Antibody [clone: 839084]

Supplier: R & D Systems
MAB7340
102887-064EA 594.37 USD
102887-064
Anti-LAMA4 Mouse Monoclonal Antibody [clone: 839084]
Antibodies
Type: Primary
Antigen: LAMA4
Clonality: Monoclonal
Clone: 839084
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR