Anti-HCRT Mouse Monoclonal Antibody [clone: 145202]

Supplier: R & D Systems
MAB734
220021-488EA 772.25 USD
220021-488
Anti-HCRT Mouse Monoclonal Antibody [clone: 145202]
Antibodies
Type: Primary
Antigen: HCRT
Clonality: Monoclonal
Clone: 145202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR