Anti-THY1 Rat Monoclonal Antibody [clone: 778053]

Supplier: R & D Systems
MAB7335
102887-062EA 594.37 USD
102887-062
Anti-THY1 Rat Monoclonal Antibody [clone: 778053]
Antibodies
Type: Primary
Antigen Symbol: THY1
Antigen Name: thymus cell antigen 1, theta
Antigen Synonyms: CD90, T25, Thy-1, Thy-1.2, Thy1.1, Thy1.2
Clonality: Monoclonal
Clone: 778053
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR